Learn More
Calcium silicate, White powder, Thermo Scientific™
For Use As a Ceramics, Paint Extender, in Chromatography
£25.12 - £125.00
Chemical Identifiers
CAS | 10101-39-0 |
---|---|
Molecular Formula | CaO3Si |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00015979 |
InChI Key | OYACROKNLOSFPA-UHFFFAOYSA-N |
Synonym | calcium metasilicate, wollastonite, aedelforsite, casiflux, cab-o-lite, gillebachite, wollastokup, calcium silicon oxide, florite, fuwalip |
PubChem CID | 26370 |
IUPAC Name | calcium;dioxido(oxo)silane |
SMILES | [O-][Si](=O)[O-].[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11453820
|
Thermo Scientific Alfa Aesar
089741.22 |
100 g | N/A | N/A | N/A | |||||
11473820
|
Thermo Scientific Alfa Aesar
089741.36 |
500 g | N/A | N/A | N/A | |||||
11463820
|
Thermo Scientific Alfa Aesar
089741.A3 |
2 kg | N/A | N/A | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
10101-39-0 | |
116.16 | |
OYACROKNLOSFPA-UHFFFAOYSA-N | |
26370 | |
[O-][Si](=O)[O-].[Ca+2] |
CaO3Si | |
MFCD00015979 | |
calcium metasilicate, wollastonite, aedelforsite, casiflux, cab-o-lite, gillebachite, wollastokup, calcium silicon oxide, florite, fuwalipShow More | |
calcium;dioxido(oxo)silane |
Specifications
1540°C | |
White | |
CaO3Si | |
calcium metasilicate, wollastonite, aedelforsite, casiflux, cab-o-lite, gillebachite, wollastokup, calcium silicon oxide, florite, fuwalip | |
OYACROKNLOSFPA-UHFFFAOYSA-N | |
calcium;dioxido(oxo)silane | |
26370 | |
Reagent | |
Calcium silicate |
10101-39-0 | |
Powder | |
MFCD00015979 | |
Insoluble in water. | |
[O-][Si](=O)[O-].[Ca+2] | |
116.16 | |
116.16 | |
2.905 |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
missing translation for 'einecsNumber' : 233-250-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.