missing translation for 'onlineSavingsMsg'
Learn More

Calcium solution for ICP 10g/L HNO3 matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Brand:  Fisher Chemical S 209 500

Code : 04

Additional Details : Weight : 0.50000kg Transport : UN number : 1760 Chem class : 8 Pack group : II


 View more versions of this product

Product Code. 12996264

  • £254.00 / 500mL
Estimated Shipment: 23-09-2024
to see stock.

Description

Description

Specifications

Specifications

10±0.02g/L
500 mL
Ca
Bottle
Mass Spectrometry Standard
HNO3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Single-element Standard Solution for ICP. Calcium (Ca) 10000mg/l in HNO3 5%

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers