Learn More
Calcium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Marque: SPEX Certiprep PLCA2-2X
Code : 04
Informations supplémentaires : CAS Number : 471-34-1 Weight : 0.50000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ca | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécification
Calcium Standard | |
Triple-leached LDPE bottle | |
94.0 | |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00010897 MFCD00085314 | |
[Ca] | |
40.08 | |
CHEBI:29320 | |
Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLCA2-2Y, -2T, -2X 10,000 PLCA2-3Y,-3X 2-5% HCl 1000μg/mL PLCA1-2X |
7440-70-2,7697-37-2,7732-18-5 | |
500 mL | |
94.0 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Ca | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
calcium | |
5460341 | |
Assurance | |
2% HNO3 |
Sécurité et traitement
- 1000
Mention d'avertissement
- Warning
Catégorie de danger
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Mention de danger
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Conseils de prudence
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
Informations supplémentaires
- MIXTURE LIST-Contains : nitric acid
Product intended for laboratory use only