missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard Metal Solution 1000ppm, for ICP, Solution HNO3 1M, Fisher Chemical

Brand:  Fisher Chemical J/8241/08


Product Code. 10090210

This item is not returnable. View return policy

Description

Description

500mL, HDPE plastic bottle

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorless
Liquid
10899
1
500 mL
HDPE plastic bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Calcium solution 1000 ppm, standard metal solution in approx. 1M nitric acid

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers