missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate, anhydrous, 99%, Thermo Scientific Chemicals

Product Code. 11379287
Click to view available options
Quantity:
250 g
1 kg
5 kg

Used as Portland-cement retarder; tile and plaster; It is the source of sulfur and sulfuric acid; Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
Synonym calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium;sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1450°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99%
Solubility Information Soluble in water,slightly soluble in glycerol.
Formula Weight 136.14
Sensitivity Hygroscopic
Appearance White powder
Density 2.96 g/cm3 at 20°C
Chemical Name or Material Calcium sulfate, anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'