missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate dihydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

£49.25 - £183.92

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium;sulfate;dihydrate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10083650
View Documents
Thermo Scientific Acros
315255000
500 g Plastic bottle
£49.25
500g
Please to purchase this item. Need a web account? Register with us today!
10572544
View Documents
Thermo Scientific Acros
315250025
2.5 kg Plastic bottle
£183.92
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10101-41-4
172.16
PASHVRUKOFIRIK-UHFFFAOYSA-L
24928
calcium;sulfate;dihydrate
CaH4O6S
MFCD00149625
calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
CHEBI:32583
O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

10101-41-4
Powder
CaH4O6S
MFCD00149625
14, 1706
Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents
O.O.[Ca++].[O-]S([O-])(=O)=O
172.16
CHEBI:32583
≥98%
Calcium sulfate dihydrate
White
98.0 to 102% (Complexometry)
CaSO4·2H2O
01,107
calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PASHVRUKOFIRIK-UHFFFAOYSA-L
calcium;sulfate;dihydrate
24928
172.17
ACS Reagent
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : EW4150000

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only