missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate Dihydrate, Extra Pure, SLR, Precipitated, Fisher Chemical
Click to view available options
500g, Plastic powder jar
Specifications
CAS | 10101-41-4 |
Color | White |
pH | 7 |
Physical Form | Powder/Solid |
Quantity | 500 g |
Molecular Formula | CaH4O6S |
MDL Number | 149625 |
InChI Key | PASHVRUKOFIRIK-UHFFFAOYSA-L |
SMILES | O.O.[Ca++].[O-]S([O-])(=O)=O |
IUPAC Name | calcium dihydrate sulfate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'