missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate Hemihydrate, Honeywell Fluka™

Product Code. 15611110
Click to view available options
Quantity:
1 kg
25 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum,≥97.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-76-1
Molecular Formula Ca2H2O9S2
Molecular Weight (g/mol) 290.283
MDL Number MFCD00799067
InChI Key ZOMBKNNSYQHRCA-UHFFFAOYSA-J
Synonym calcium sulfate hemihydrate, gypsum hemihydrate, calcium sulphate hemihydrate, unii-3rw091j48v, crystacal, densite, hemihydrate gypsum, densite gypsum, gipshalbhydrat, p 23 sulfate
PubChem CID 3033839
ChEBI CHEBI:32584
IUPAC Name dicalcium;disulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ca+2].[Ca+2]

Specifications

Quantity 1 kg
Linear Formula CaSO4 · 0.5H2O
UN Number NONH for all modes of transport
Formula Weight 145.15g/mol
Percent Purity ≥97%
Grade purum
Chemical Name or Material Calcium Sulfate Hemihydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'