missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate, Technical, Anhydrous, Powder, SLR, Fisher Chemical™
Chemical Identifiers
CAS | 7778-18-9 |
---|---|
Molecular Formula | CaO4S |
Molecular Weight (g/mol) | 136.13 |
MDL Number | MFCD00010912 |
InChI Key | OSGAYBCDTDRGGQ-UHFFFAOYSA-L |
IUPAC Name | calcium sulfate |
SMILES | [Ca++].[O-]S([O-])(=O)=O |
Description
Calcium Sulfate, Technical, Anhydrous, Powder, SLR
Chemical Identifiers
7778-18-9 | |
136.13 | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
[Ca++].[O-]S([O-])(=O)=O |
CaO4S | |
MFCD00010912 | |
calcium sulfate |
Specifications
1450°C | |
White | |
CaO4S | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
calcium sulfate | |
136.14g/mol |
7778-18-9 | |
Solid | |
MFCD00010912 | |
[Ca++].[O-]S([O-])(=O)=O | |
136.13 | |
Plastic powder jar |