missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate, Technical, (Plaster Of Paris), Fisher Chemical

Product Code. 10620331 Shop All Fisher Chemical Products
Click to view available options
:
3kg, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10620331

Brand: Fisher Chemical C/2280/63

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 1450°C
CAS 7778-18-9
Color White
pH 6 to 7.6
Physical Form Solid
Quantity 3 kg
Molecular Formula CaO4S
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES [Ca++].[O-]S([O-])(=O)=O
IUPAC Name calcium sulfate
Molecular Weight (g/mol) 136.13
Formula Weight 145.15g/mol
Packaging Plastic powder jar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.