missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfide, 99% (metals basis excluding Mg), Mg <1%, Thermo Scientific Chemicals

£178.89 - £532.65

Chemical Identifiers

CAS 20548-54-3
Molecular Formula CaS
Molecular Weight (g/mol) 72.14
MDL Number MFCD00015990
InChI Key AGVJBLHVMNHENQ-UHFFFAOYSA-N
PubChem CID 30182
IUPAC Name calcium sulfanediide
SMILES [S--].[Ca++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11335867
View Documents
Thermo Scientific Alfa Aesar
043646.14
25 g
£178.89
25g
Please to purchase this item. Need a web account? Register with us today!
11345867
View Documents
Thermo Scientific Alfa Aesar
043646.22
100 g
£532.65
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcium sulfide is used in phosphors, lubricant additives and luminous paints. It reacts with elemental sulfur to give a lime-sulfur. Further, it is used in ore dressing and flotation agent. In addition to this, it is used as a depilatory in the tanning industry and in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20548-54-3
72.14
AGVJBLHVMNHENQ-UHFFFAOYSA-N
calcium sulfanediide
CaS
MFCD00015990
30182
[S--].[Ca++]

Specifications

(decomposition)
White
CaS
UN3134
Slightly soluble in water. Insoluble in alcohol.
[S--].[Ca++]
72.14
72.14
Stench
2.5 g/mL
Calcium sulfide
20548-54-3
Powder
MFCD00015990
14,1707
AGVJBLHVMNHENQ-UHFFFAOYSA-N
calcium sulfanediide
30182
99%
Moisture sensitive
(metals basis excluding Mg), Mg <1%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335-H500

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II

missing translation for 'einecsNumber' : 243-873-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only