missing translation for 'onlineSavingsMsg'
Learn More

Calcium, Technical, Metal, Pieces, SLR, Fisher Chemical™

Product Code. 11443663 Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
This item is not returnable. View return policy

Product Code. 11443663

Brand: Fisher Chemical C/0840/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ca, CAS Number-7440-70-2, 100g, 1484 deg.C, Gray, 40.07g/mol, 845 deg.C, 85314, Plastic powder jar, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
IUPAC Name calcium
SMILES [Ca]

Specifications

Boiling Point 1484°C
Melting Point 845°C
Color Gray
pH 14
Physical Form Solid
Quantity 250 g
Formula Weight 40.07g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Calcium
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P223-Do not allow contact with water.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.