missing translation for 'onlineSavingsMsg'
Learn More
Calcium tetrafluoroborate hydrate, Thermo Scientific Chemicals Product Code.: 11312878

Calcium tetrafluoroborate hydrate, Thermo Scientific Chemicals

Product Code. 11312878
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11312878

Brand: Thermo Scientific Alfa Aesar 030131.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calcium tetrafluoroborate hydrate is used as pharmaceutical and chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13814-93-2
Molecular Formula B2CaF8
Molecular Weight (g/mol) 213.685
MDL Number MFCD00015972
InChI Key YHLNCCJTKRTQEG-UHFFFAOYSA-N
Synonym calcium tetrafluoroborate, calcium ditetrafluoroborate, calcium tetrafluoroborate 1-, acmc-1bo7f
PubChem CID 6096284
IUPAC Name calcium;ditetrafluoroborate
SMILES [B-](F)(F)(F)F.[B-](F)(F)(F)F.[Ca+2]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 213.68 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Calcium tetrafluoroborate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium tetrafluoroborate hydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.