missing translation for 'onlineSavingsMsg'
Learn More

Calcon™ Indicator, For Complexometry, Honeywell Fluka™

Product Code. 15605570
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator, for complexometry (Al, Fe, Zr)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2538-85-4
Molecular Formula C20H13N2NaO5S
Molecular Weight (g/mol) 416.383
MDL Number MFCD00004069
InChI Key YJUGDMOGPJJWDG-UHFFFAOYSA-M
PubChem CID 23702480
IUPAC Name sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+]

Specifications

UN Number NONH for all modes of transport
Beilstein 6041435
Solubility Information Method: 70mg in 10mL N,N-Dimethylformamide
Formula Weight 416.38g/mol
Packaging Plastic Bottle
Color Brown
Quantity 100 g
Physical Form Powder
Chemical Name or Material Calcon™
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'