missing translation for 'onlineSavingsMsg'
Learn More

Calconcarboxylic Acid, Honeywell Fluka™

Product Code. 15681940 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 15681940

Brand: Honeywell-Fluka 2126010G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for complexometry

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3737-95-9
Molecular Formula C21H14N2O7S
Molecular Weight (g/mol) 438.41
MDL Number MFCD00004078
InChI Key ULIVOAKVRBXKKS-PYCFMQQDSA-N
PubChem CID 5895210
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)O)O)C(=O)O

Specifications

Melting Point 300°C
UN Number NONH for all modes of transport
Beilstein 727492
Formula Weight 438.41g/mol
Packaging Plastic Bottle
Color Violet
Quantity 10 g
Physical Form Powder
Chemical Name or Material Calconcarboxylic Acid, For complexometry
compliance-icons
Product Identifier
  • Calconcarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,2'-Dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.