missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Calconcarboxylic acid, indicator grade Product Code.: 10501411

Thermo Scientific Chemicals Calconcarboxylic acid, indicator grade

Product Code. 10501411
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10501411

Brand: Thermo Scientific Chemicals 157310100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calconcarboxylic acid, indicator grade, Acros Organics, Quantity: 250g, Packaging: Glass bottle, Black to Purple, Grade: Extra Pure, Melting Point: 300 deg.C, Molecular Weight: 438.4, CAS: 3737-95-9, EINECS Number: 223-117-, Formula Weight: 438.4, Health Hazard 1: GHS Signal Word: Warning

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3737-95-9
Molecular Formula C21H14N2O7S
Molecular Weight (g/mol) 438.4
MDL Number MFCD00004078
InChI Key ULIVOAKVRBXKKS-PYCFMQQDSA-N
Synonym calconcarboxylic acid, patton-reeder indicator, calconcarbonic acid, calcon 3-carboxylic acid, patton and reeder's indicator, 2-hydroxy-1-2-hydroxy-4-sulfo-1-napthylazo-3-naphthoic acid, 2,2'-dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy-4-2-hydroxy-4-sulfo-1-naphthalenyl azo, nnShow MoreShow Less
PubChem CID 5895210
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)O)O)C(=O)O

Specifications

Melting Point 300°C
Color Black to Purple
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol and ethanol
Formula Weight 438.4
Grade Extra Pure
Physical Form Powder
Chemical Name or Material Calconcarboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464929 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number157310500,157312500,157310100
Lot NumberA0458048 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number157310500,157312500,157310100
Lot NumberA0397566 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number157310500,157312500,157310100
Lot NumberA0393831 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number157310500,157312500,157310100
Lot NumberA0395606 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number157310500,157312500,157310100
compliance-icons
Product Identifier
  • Calconcarboxylic acid, indicator grade
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    1

    Hours

    13

    Minutes

    7

    Seconds

    20

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Calconcarboxylic acid, indicator grade >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.