missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcyphosine Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | Calcyphosine |
---|---|
Concentration | 0.26 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836704
|
Proteintech
16926-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16826704
|
Proteintech
16926-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Calcyphosine | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q13938 | |
828 | |
Calcyphosine Fusion Protein Ag10330 | |
Primary | |
-20°C | |
CAPS |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Calcyphosin, calcyphosine, CAPS, CAPS1 | |
CAPS | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |