missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 7.01 Calibration Buffer Sachets, Hanna Instruments™
missing translation for 'orderingAttributeHoverText'
Quantity:
20 x 25 mL
missing translation for 'unitSize'
Pack of 25
Description
- Premium quality pH 7.01 calibration buffer solution produced in accordance with ISO 3696/BS3978 standards
- Color coded label for easy pH buffer value identification
- Temperature chart of the actual pH value at various temperatures printed on each sachet
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
- Ideal for users that do not calibrate frequently
- Ideal for field use due to portability
Specifications
Specifications
| pH | 7.01 |
| Physical Form | Liquid |
| Quantity | 20 x 25 mL |
| Packaging | Sachets |
| Traceability to NIST | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction