missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calibration Buffer Solutions, Hanna Instruments™
Click to view available options
Quantity:
20 mL
pH:
4.01
10.01
Description
- Premium quality pH 10.01 calibration buffer solution
- 25 sachets containing 20 mL of buffer
- Color coded label for easy pH buffer value identification
- Accuracy of +/- 0.01 pH @ 25°C
- Temperature chart of the actual pH value at various temperatures printed on each sachet
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware
- In a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
- Ideal for users that do not calibrate frequently
- Ideal for field use due to portability
Specifications
Specifications
Color | Colorless |
pH | 10.01 |
Physical Form | Liquid |
Quantity | 20 mL |
Solution Type | Buffer |
Formulation | Aqueous Buffer Solution |
Odor | Odorless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction