Learn More
BINDER™ Calibration Certificate 18 P Temperature for Series KBF-S, KBF-S ECO
BINDER™ Calibration certificate is an option for use with Constant Climate Chambers KBF-S and KBF-S ECO
Brand: BINDER™ 8012-2068
Description
- Calibration certificate, temperature
- Temperature measurement
- Includes certificate
- 15 to 18 measuring points at specified temperature
Specifications
BINDER Chambers Series KBF-S ECO | |
60 to 120 min. | |
Certificate 15- 18 measuring points (at specified temperature) |
Calibration certificate, temperature - temperature measurement incl. certificate, 15- 18 measuring points (at specified temperature) | |
Calibration Certificate |
Product Suggestions
Customers who viewed this item also viewed