missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Calibration Certificate, Temperature and Humidity
Calibration certificate, temperature and humidity - Measurement in center of chamber at 25°C / 60% RH or at specified test values
£475.00 - £2710.00
Specifications
Product Type | Calibration Certificate (Temperature and Humidity) |
---|---|
For Use With (Application) | Temperature and Humidity Measurement |
Product Code | Brand | For Use With (Equipment) | Calibration Points | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Calibration Points | Price | Quantity & Availability | ||||
15922962
|
BINDER™
8012-1187 |
BINDER Chambers (Models MKFT 115, MKFT 240, MKFT 720) | 1 measuring point |
£475.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932962
|
BINDER™
8012-1188 |
BINDER Chambers (Models MKF 56, MKF 115, MKF 240, MKF 720) | 1 measuring point |
£475.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933012
|
BINDER™
8012-1614 |
BINDER Chambers (Models MKFT 115, MKFT 240, MKFT 720) | 27 measuring points |
£2710.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943012
|
BINDER™
8012-1615 |
BINDER Chambers (Models MKF 56, MKF 115, MKF 240, MKF 720) | 27 measuring points |
£2710.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- One humidity point of measurement
- Including certificate
- Humidity measurement point in center of chamber
- Temperature (according to DIN12880) and humidity measurement at 25°C / 60% RH or at specified values
Specifications
Calibration Certificate (Temperature and Humidity) | |
Temperature and Humidity Measurement |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Calibration Certificate, Temperature and Humidity