Learn More
Velp Scientifica™ Calibration Set
Velp Scientifica™ Calibration Set for use with TB1 Portable Turbidimeter.
Brand:  Velp Scientifica™ CE0012020
Product Code. 15379644
  • £435.00 / Each
Explore more special offers
Description
Description
  • For use with TB1 Portable Turbidimeter
Specifications
Specifications
Calibration Kit
TB1 Turbidimeter
800, 100, 20.0 & 0.02 NTU standard
Measures the turbidity of aqueous samples simply and accurately
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Velp Scientifica™ Calibration Set > Product Type: Calibration Set