missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calibration Solution, Hanna Instruments™
Click to view available options
pH:
4.01
7.01
Description
- Premium quality
- Lot number and expiration date clearly marked on label
- Air tight with tamper-proof seal to ensure quality of solution NIST traceable and supplied with Certificate of Analysis
- Easy to identify different buffer values
- Solution does not contain any dyes that may stain reference cell of pH electrode
Specifications
Color | Undesignated |
Certificate of Analysis | Yes |
pH | 7.01 |
Quantity | 500 mL |
Traceability to NIST | Yes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'