Learn More
Thermo Scientific™ Calibration Validation Carousels
Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money.
£5345.00 - £7025.00
Specifications
Certifications/Compliance | All standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom. |
---|---|
Description | NIST traceable |
For Use With (Equipment) | Evolution 201, 220, 260 Bio and 300 |
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
11556959
|
Thermo Scientific™
10010721 |
Routine Performance Validation - NPL Traceable Filters | Calibration Validation Carousel with Smart Base |
£5345.00
Each |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10031518
|
Thermo Scientific™
840-213000 |
Pharmacopia Peformance Validation - EP | Calibration Validation Carousel with Smart Base |
£7025.00
Each |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money. The CVC provides certified reference materials in a fully automated confirguration for hands-free performance verification of your Thermo Scientific™ spectrophotometer.
- Simplify qualification procedures by automating performance verification of UV-Visible spectrophotometers
- Each CVC has a unique serial number matched automatically to the corresponding data files containing standard values and instrument specifications
- Generated result reports are suitable for printing or archival purposes
- Included standards are traceable to either NIST™ primary standards or fundamental physical standards
- CVCs are available in standard or pharmacopeia compliant configurations
Comprehensive Performance Verification Testing
- Wavelength Accuracy and Repeatability
- Photometric Accuracy and Repeatability
- Stray Light
- Resolution
- Photometric Noise
- Photometric Stability or Drift
- Baseline Flatness
Save Time, Reduce Errors
- Eliminates need to copy standard values from certificates
- Eliminates need to analyze standards individually
- Eliminates need to match standards to data
- Eliminates need for manual calculations
- Eliminates need for manual transcription and reporting of results
Ordering Information:
Unit is shipped in a temperature-protected plastic case for optimal protection during storage. Retain this case and packaging to use when sending the unit for periodic recalibration. Calibration Validation Carousels can be used with Thermo Scientific™ INSIGHT™ and INSIGHT Security, Thermo Scientific™ VISIONpro™ and VISIONsecurity™. Pharmacopeia-compliant CVCs are compatible with the Thermo Scientific™ Evolution 300, 260 Bio, 220 and 201 UV-Visible spectrophotometers.
Specifications
All standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom. | |
NIST traceable | |
Evolution 201, 220, 260 Bio and 300 |