missing translation for 'onlineSavingsMsg'
Learn More

Calmidazolium chloride, Tocris Bioscience™

Product Code. 18054302
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Calmodulin antagonist

Calmodulin antagonist. Inhibits calmodulin-dependent phosphodiesterase and Ca2+-transporting ATPase with IC50 values of 0.15 and 0.35 μM respectively. Also causes elevation of intracellular calcium in HL-60 cells, independent of calmodulin inhibition.

Specifications

CAS 57265-65-3
Molecular Formula C31H23Cl7N2O
Quantity 10 mg
Synonym calmidazolium chloride, 1-bis 4-chlorophenyl methyl-3-2-2,4-dichlorophenyl-2-2,4-dichlorobenzyloxy ethyl-1h-imidazolium chloride, 3-bis 4-chlorophenyl methyl-1-2-2,4-dichlorophenyl-2-2,4-dichlorophenyl methoxy ethyl imidazol-1-ium chloride, r 24571 chloride, methoxy ethyl-, chloride, calmidazolium chloride, solid, 1-bis p-chlorophenyl methyl-3-2,4-dichloro-beta-2,4-dichlorobenzyloxy phenethyl, 1-bis 4-chlorophenyl methyl-3-2,4-dichloro-, a-2,4-dichlorobenzyloxy phenethyl imidazolium chloride, 1-bis 4-chlorophenyl methyl-3-2,4-dichloro-beta-2,4-dichlorobenzyloxy phenethyl imidazolium chloride, 1-bis 4-chlorophenyl methyl-3-2-2,4-dichlorophenyl-2-2,4-dichlorophenyl methoxy ethyl imidazol-1-ium chloride
InChI Key YGEIMSMISRCBFF-UHFFFAOYSA-M
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)N3C=C[N+](=C3)CC(C4=C(C=C(C=C4)Cl)Cl)OCC5=C(C=C(C=C5)Cl)Cl)Cl.[Cl-]
IUPAC Name 1-[bis(4-chlorophenyl)methyl]-3-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazol-3-ium;chloride
Molecular Weight (g/mol) 687.688
PubChem CID 644274
Formula Weight 687.7
Percent Purity >98%
Chemical Name or Material Calmidazolium chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'