Learn More
Thermo Scientific Chemicals Caloxin 2A1
For cellular and molecular biology applications
Brand: Thermo Scientific Chemicals J66428.M¬
Code : NEW
Additional Details : CAS Number : 350670-85-8 Weight : 0.00001kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
Caloxin 2A1 | |
RUO - research use only |
-30°C to -10°C | |
1478.54 | |
1 mg | |
Val-Ser-Asn-Ser-Asn-Trp-Pro-Ser-Phe-Pro-Ser-Ser-Gly-Gly-Gly-NH2 |
RUO – Research Use Only