Learn More
Calreticulin Recombinant Rabbit Monoclonal Antibody (SU37-03), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA532131
301.70 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
Specifications
Calreticulin | |
Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide, pH 7.4 | |
P14211, P18418, P27797 | |
CALR | |
Synthetic peptide within Human Calreticulin aa 40-89 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry, Western Blot | |
SU37-03 | |
Unconjugated | |
CALR | |
Autoantigen RO, CABP3, CALBP, calcium-binding protein 3, CALR, calrectulin, calregulin, Calreticulin, calreticulin precursor, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, I79_008346, RO, Ro/SS-A Antigen, Sicca syndrome antigen A (autoantigen Ro, calreticulin), SSA | |
Rabbit | |
Protein A | |
RUO | |
12317, 64202, 811 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.