missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cambendazole, Honeywell Fluka™
Click to view available options
Quantity:
10 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Powder |
CAS | 26097-80-3 |
Molecular Formula | C14H14N4O2S |
MDL Number | MFCD00864552 |
Beilstein | 563292 |
Molecular Weight (g/mol) | 302.35 |
Grade | Analytical Standard |
Packaging | Glass Bottle |
Quantity | 10 mg |
For Use With (Application) | Anthelmintics and Flukicides |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'