missing translation for 'onlineSavingsMsg'
Learn More

Carl Zeiss™ Camera Adapters

Product Code. 15150705
Click to view available options
Description:
60N-C 0.75 in.
60N-C 1.0 in.
Magnification Power:
0.5x
0.63x
1.0X
Unit Size:
1 unit
Each
This item is not returnable. View return policy

Product Code. 15150705

Brand: Carl Zeiss™ 4261140000000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

C-mount camera adapter for Primovert

  • Requires Primovert
  • With a phototube C mount camera adapter
  • For Primovert

Specifications

Type Camera Adapter
Description 60N-C 1.0 in.
Magnification Power 1.0X
Quantity 1 Ea.
Size 1.0 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.