missing translation for 'onlineSavingsMsg'
Learn More

CaMKII alpha Monoclonal Antibody (6G9), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA1048

Product Code. 11528772

  • £449.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MA1-048 detects CaM kinase II from rat and mouse tissues. This antibody has been shown to detect both the phosphorylated and non-phosphorylated alpha subunit of CaM kinase II only. MA1-048 does not detect the ∽60 kDa beta subunit in either phosphorylation state. MA1-048 has been successfully used in Western blot, immunohistochemistry, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects an ∽50 kDa protein representing the alpha subunit of CaM kinase II. Immunofluorescence staining of CaM kinase II in rat hippocampal cells with MA1-048 results in intense staining of cell somas and neuropil areas, while nuclei are only lightly stained. The MA1-048 immunogen is partially purified, rat CaM kinase II.

CAMK2A (CaMKII alpha) is a serine/threonine kinase that regulates a number of cellular functions including neuronal processes, gene expression, carbohydrate metabolism, cytoskeletal function, and calcium homeostatsis. Autophosphorylation of threonine 286 in the presence of Ca2+ and calmodulin activates CAMK2A with the ability to switch from calmodulin-dependent to a calmodulin independent activity. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CaMKII alpha
Monoclonal
1.17 mg/mL
PBS with 0.05% sodium azide
P11275, P11798
CAMK2A
Partially purified, rat CaM kinase II.
100 μL
Primary
Mouse, Rat
Antibody
IgG1
Immunohistochemistry, Immunoprecipitation, Western Blot
6G9
Unconjugated
CAMK2A
alpha CaM kinase II; alpha-CaMKII; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; Calci; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; CaM kinase II alpha subunit; caM kinase II subunit alpha; CaMK II; CAMK2; Camk2a; CAMKA; CaMKII; CaMK-II alpha subunit; caMK-II subunit alpha; CaMKIINalpha; CaM-kinase II alpha chain; EC 2.7.11.17; KIAA0968; mKIAA0968; OTTHUMP00000165787; OTTHUMP00000165788; Phospho-CaMKI; PK2CDD; PKCCD; R74975
Mouse
Protein A
RUO
12322, 25400
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers