missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMKV Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14788-1-AP-150UL
This item is not returnable.
View return policy
Description
CaMKV does not appear to have detectable kinase activity.Specifications
CAMKV | |
Polyclonal | |
Unconjugated | |
CAMKV | |
1G5, CAMKV, VACAMKL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235604, 79011, 79012 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UHL1, Q63092, Q8NCB2 | |
Camkv | |
CAMKV Fusion Protein Ag6438 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |