missing translation for 'onlineSavingsMsg'
Learn More

Cytiva cAMP Direct BioTrak™ EIA

Use for direct measurement of cAMP in cell lysates without any need for removal of extraction reagents. Cytiva Life Sciences™ cAMP Direct BioTrak™ EIA represents a convenient, high-sensitivity, peroxidase-based technology that enables the amount of protein present in a sample to be measured in a single day.

Brand:  Cytiva RPN2251


 View more versions of this product

Product Code. 10451835

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Flexible method-choice of assay protocols
  • Rapid assay protocol
  • Non-radioactive
  • Precise and accurate measurement
  • Ready-to-use substrate
  • Color coded reagents
  • Five-hour protocol

  • Kits available with or without acetylation reagents
  • Bulk kit provided without lysis buffers
  • Dual range: 25–6400 fmol/well [non-acetylation], 2–128 fmol/well [acetylation]
  • Sensitivity: 38.4pg/mL [non-acetylation] and 14pg/mL [acetylation]

TRUSTED_SUSTAINABILITY
Specifications

Specifications

38.4pg/mL (non-acetylation)
96-well Microplate
96-well coated plates, Acetylation and non-acetylation assay standard lysis reagent 1, Antiserum, Assay buffer, Lysis reagent 2, Peroxidase conjugate cAMP-HRP, lyophilized, TMB substrate, Wash buffer
2°C to 8°C
cAMP biotrak Enzyme immunoassay (EIA) system
Used for research purposes, eliminates lengthy sample extraction procedures and allow direct cAMP measurement
96 Determinations
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P270-Do not eat, drink or smoke when using this product.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P264-Wash hands thoroughly after handling.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P405-Store locked up.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : calcium chloride
  • UNKNOWN ACUTE-100 (kit) per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100 (kit)% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers