missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Camphor-10-sulfonic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
(+)-Camphor-10-sulfonic acid is used as a resolving agent for chiral amines and other cations. It is involved in the preparation of active pharmaceutical ingredient such as trimetaphan camsilate, which is used to reduce bleeding during neurosurgery. Further, it is involved in the transglucosidation of methyl and ethyl D-glucopyranosides by alcoholysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+)-Camphor-10-sulfonic acid is used as a resolving agent for chiral amines and other cations. It is involved in the preparation of active pharmaceutical ingredient such as trimetaphan camsilate, which is used to reduce bleeding during neurosurgery. Further, it is involved in the transglucosidation of methyl and ethyl D-glucopyranosides by alcoholysis.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
(+)-Camphor-10-sulfonic acid is used as a resolving agent for chiral amines and other cations. It is involved in the preparation of active pharmaceutical ingredient such as trimetaphan camsilate, which is used to reduce bleeding during neurosurgery. Further, it is involved in the transglucosidation of methyl and ethyl D-glucopyranosides by alcoholysis.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼200°C (decomposition) |
Quantity | 100 g |
UN Number | UN3261 |
Beilstein | 3205973 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 232.3 |
Percent Purity | 98% |
Chemical Name or Material | (+/-)-Camphor-10-sulfonic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'