missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Camptothecin

Product Code. p-7082313
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15484889

Brand: Thermo Scientific Chemicals J62523.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cytotoxic antitumor agent that inhibits topoisomerase I | CAS: 7689-03-4 | C20H16N2O4

Camptothecin is used as a cytotoxic antitumor agent that inhibits topoisomerase I.Camptothecin shows remarkable anticancer activity based on preliminary clinical trials. Its analogues are used in cancer chemotherapy. Cyclodextrin-based polymer containing camptothecin is used in the investigational anti-cancer drug study.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-4-7689
Synonym (S)-(+)-Camptothecin

Specifications

Description Yellow
Quantity 250 mg
Molecular Formula C20H16N2O4
Formula Weight 348.35g/mol
Melting Point 260°C
Product Type Apoptosis Reagent
Content And Storage 2°C to 8°C
For Use With (Application) Cytotoxic antitumor agent that inhibits topoisomerase I
Form Powder
compliance-icons
Product Identifier
  • Camptothecin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : Camptothecin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.