missing translation for 'onlineSavingsMsg'
Learn More

Novacyt - Microgen Bioproducts Campylobacter Latex Agglutination Test

Product Code. 17313191 Shop All Novacyt Group Products
Click to view available options
No. of Reactions:
2 per test
Unit Size:
50 tests
This item is not returnable. View return policy

Product Code. 17313191

Brand: Novacyt - Microgen Bioproducts F46

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Microgen Bioproducts Campylobacter - A rapid latex slide agglutination test for enteropathogenic Campylobacter grown in enrichment broths or on selective plates.

  • Able to detect the presence of Campylobacter spp. in clinical and food samples.

Specifications

Certifications/Compliance ISO13485 ISO9001
Content And Storage 2°C to 8°C
Control Sets F46c Positive Control - 1.0 mL
CE Marker No
Clia Complexity Non-waived
Detectable Analytes C. jejuni Antibodies (Rabbit)
Format 4 Bottles (2.5 mL, 2.5 mL, 1.0 mL, 5.0 mL)
Sample Type Animal, avian, food or environmental origin
Sensitivity 0.986
Type Latex Agglutination Test
For Use With (Application) Confirmatory Identification of Thermophilic Campylobacter Spp. (C. Jejuni, C. Coli, C. Jejuni Subsp. Doylei, C. Lari, C. Upsaliensis and C. Fetus) Cultured on Selective Solid Media
For Use With (Equipment) Bacteriological Loops, Disposable Plastic Specimen Tubes (Capacity 1 mL), Apparatus for Maintaining Microaerophilic Culture Conditions, 42°C Incubator (if Not Available, a 37°C Incubator is Acceptable)
Includes 50 Tests
Material Polythene Bottle
No. of Reactions 2 per test
Organism Group Animal/plant
Packaging Type Carton with Insert vial box
Quantity 4 Bottles (2.5 mL, 2.5 mL, 1.0 mL, 5.0 mL)
Reagent Type F46a Test Latex Reagent - 2.5 mL Latex particles coated with rabbit antibodies to Campylobacter antigens
Species Animal, avian, food or environmental, not human
Specificity 0.997
Target Organism Class Eukaryota
Test Time 2 Minutes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato