Learn More
Thermo Scientific™ Campylobacter Selective Agar (Skirrow)
Description
Selectively isolate Campylobacter jejuni and Campylobacter coli from human, animal, avian and environmental specimens with Thermo Scientific™ Oxoid™ Campylobacter Selective Agar (Skirrow). The medium is recommended for the isolation of Campylobacter jejuni and Campylobacter coli from fecal samples. The medium was devised to overcome the practical difficulties associated with the fecal filtrate culture technique described by Butzler1. Furthermore for optimal selectivity Campylobacter Selective Agar (Skirrow) can be incubated at 42°C.
Isolate campylobacter species from fecal samples using Campylobacter Selective Agar (Skirrow). The medium is a modification of the formulation described by Skirrow2
- Can be optimized for better selectivity by incubating at 42°C.
- Helps avoid the practical difficulties associated with the fecal filtrate culture technique.
- Ready-to-use convenience of prepared media
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Color | Red / Brown translucent |
| Content And Storage | 2°C to 10°C |
| Description | Campylobacter Selective Agar (Skirrow) |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 19.5g±1.0.g |
| Packaging Type | 10 x 90mm plates, film wrapped |
| Quantity | 10/Pk. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.