Learn More
Cancer-Targeting Compound 1, MedChemExpress
MedChemExpress Cancer-Targeting Compound 1 is used in the research of hormone-related cancer, extracted from patent WO 2008021331 A2.
Brand: Medchem Express HY-U00300
Description
Cancer-Targeting Compound 1 is used in the research of hormone-related cancer, extracted from patent WO 2008021331 A2.Specifications
Cancer-Targeting Compound 1 | |
White | |
C15H13N3O2S | |
N#CC1=CC=C(C2=CSC(N3C(OCC3(C)C)=O)=N2)C=C1 | |
299.35 | |
Research | |
Cancer-programmed cell death |
1007581-62-5 | |
Research | |
1 mg | |
299.35 | |
98.37% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed