Learn More
Candesartan-d4, MedChemExpress
MedChemExpress Candesartan D4 (CV-11974 D4) is the deuterium labeled Candesartan, which is an angiotensin II receptor antagonist.
£400.51 - £1203.95
Specifications
Chemical Name or Material | Candesartan-d4 |
---|---|
CAS | 1346604-70-3 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C24H16D4N6O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16480818
|
Medchem Express
HY-B0205S |
5 mg |
£802.23
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16468418
|
Medchem Express
HY-B0205S |
10 mg |
£1203.95
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16412089
|
Medchem Express
HY-B0205S |
1 mg |
£400.51
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Candesartan D4 (CV-11974 D4) is the deuterium labeled Candesartan, which is an angiotensin II receptor antagonist.Specifications
Candesartan-d4 | |
White | |
C24H16D4N6O3 | |
[2H]C1=C(CN2C(C(C(O)=O)=CC=C3)=C3N=C2OCC)C([2H])=C([2H])C(C4=CC=CC=C4C5=NNN=N5)=C1[2H] | |
444.48 | |
Research |
1346604-70-3 | |
Research | |
CV-11974-d4 | |
444.48 | |
98.85% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.