missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Candida albicans Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA127158
This item is not returnable.
View return policy
Description
PA1-27158 detects C. albicans and other yeasts from yeast samples.
Candida albicans is the most frequently isolated fungal pathogen of humans, affecting immuocompromised patients ranging from premature infants to AIDS sufferers. Systemic infections have an attributed mortality of 30-50%. C. albicans is a diploid organism which has eight sets of homologous chromosomes. Its genome size is about 16 Mb (haploid), about 30% greater than S. cerevisiae (baker's yeast).Specifications
Candida albicans | |
Polyclonal | |
Unconjugated | |
C. albicans | |
Candida albicans, type A. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunodiffusion, Immunoelectrophoresis, Immunohistochemistry | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
IgG fraction | |
RUO | |
Yeast | |
Antibody | |
IgG |