missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Canine IL-10 DuoSet ELISA
Canine IL-10 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DY735
Additional Details : Weight : 0.09700kg
Description
The Canine IL-10 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies canine IL-10.Specifications
Biotin | |
ELISA | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF |
31.20 to 2000 pg/mL | |
403628 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |