Learn More
Thermo Scientific™ ABgene™ Cap strip, for PCR tubes, strips of eight flat caps
Brand: Thermo Scientific™ ABgene™ AB-0784-RMP340
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Compatible with 96-well PCR plates and standard and low profile 0.2mL strip tubes
- Certified DNase, RNase and human DNA free
- Domed and flat available
- Ultra clear model available for QPCR and fluorescence applications
- Compatible with 96-well PCR plates and standard and low profile strip tubes