missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Capillaries for Melting Point Apparatus

For use with BUCHI Melting Point apparatus.

£52.30 - £253.00

Specifications

Product Type Melting Point Capillary
Dimensions (L x W x H) 180 x 120 x 80 mm
For Use With (Equipment) BUCHI Melting Point Apparatus
Height (Metric) 80 mm
Material Glass
View More Specs

Products 2
Product Code Brand Packaging Quantity Price Quantity & Availability  
Product Code Brand Packaging Quantity Price Quantity & Availability  
10421161
View Documents
BUCHI
001759
1000 Tubes
£253.00
Pack of 1000
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
10108981
View Documents
BUCHI
017808
100 Tubes
£52.30
Pack of 100
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
Description

Description

  • High-quality Borosilicate 3.3 glass
  • Optimized thickness
  • Reliable results
  • Compatible with other automated instrument or manual techniques
Specifications

Specifications

Melting Point Capillary
BUCHI Melting Point Apparatus
Glass
180 x 120 x 80 mm
80 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BUCHI Capillaries for Melting Point Apparatus

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.