Learn More
Applied Biosystems™ Capillary Cutter, 310 Genetic Analyzer
This is a capillary cutter for use with 310 genetic analyzer capillaries. Shortened capillaries can be used for short run modules.
Brand:  Applied Biosystems™ 401958
Additional Details : Weight : 0.00450kg
Product Code. 17142959
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Store at room temperature.
Sequencing
2 Each
310 Genetic Analyzer
Capillary Cutter
Room Temperature