Learn More
Thermo Scientific™ Capitol Vial 10mL and 12mL Screw Top Transport Tubes and Caps
Choose Capitol Vial 10 mL and 12 mL screw top tubes for sample collection and transport. The tubes are manufactured from a food grade polypropylene resin for visual clarity and sample security.
Brand: Thermo Scientific™ S40100
148.04 GBP valid until 2024-12-31
Use promo code "21960" to get your promotional price.
Additional Details : Weight : 0.01100kg
Description
Thermo Scientific™ Capitol Vial 10mL and 12mL Screw Top Transport Tubes are made from virgin, clarified polypropylene resin for visual clarity and sample security.
Features:
- Freestanding
- Nonsterile tubes
10 mL:
- Graduations in 1 mL increments up to 8 mL
- 0.59 O.D. x 3.38 in. L (15 x 86 mm)
- Round bottom with skirt
- Primary application: serum transport
12 mL:
- Etched fill lines at 5 mL and 8 mL
- 0.59 O.D. x 3.77 in. L (15 x 96 mm)
- Conical bottom with skirt
- Available in amber color for light-sensitive samples
- For general transport
Specifications
10mL transport tube; Clear; 2000/Cs. | |
Polypropylene | |
10 mL | |
Skirted, round bottom | |
86 mm | |
Capitol Vial Screw Caps (not included) |
Clear | |
Non-sterile | |
2000 | |
Screw Cap | |
Sample Tube |