missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Capitol Vial 10mL and 12mL Screw Top Transport Tubes and Caps
GREENER_CHOICE

Choose Capitol Vial 10 mL and 12 mL screw top tubes for sample collection and transport. The tubes are manufactured from a food grade polypropylene resin for visual clarity and sample security.

Brand:  Thermo Scientific™ S40204

92.39 GBP valid until 2024-12-31
Use promo code "21960" to get your promotional price.


 View more versions of this product

Product Code. 17145311

  • £106.00 / Pack of 2000
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Capitol Vial 10mL and 12mL Screw Top Transport Tubes are made from virgin, clarified polypropylene resin for visual clarity and sample security.

Features:

  • For 10 and 12 mL serum transport tubes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polyethylene
Yellow
2000/Cs.
10 and 12 mL Serum Transport Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers