Learn More
Thermo Scientific™ Capitol Vial™ 45 mL Flip-Top Polypropylene Container
Thermo Scientific™ Capitol Vial™ 45 mL Flip-Top Polypropylene Containers are ideal for the transport of urine samples for drug testing programs. The containers feature tamper evident and locking seals for sample security before and after collection.
Brand: Thermo Scientific™ 10153
Additional Details : Weight : 10.45000kg
Description
Thermo Scientific™ Capitol Vial™ 45 mL Flip-Top Polypropylene Containers are ideal for the transport of urine samples for drug testing programs. The containers feature tamper evident and locking seals for sample security before and after collection.
- Air tight and leak resistant construction
- Food-grade polypropylene
- Permanent hinge lid allows easy one-handed opening of vial
- Shoulderless, liner-less containers
Drugs of abuse testing, food and beverage sample collection, veterinary applications, environmental sampling, water testing, general packaging and storage
Specifications
45 mL Flip Top Vial, Twin Valve | |
Non-sterile | |
700 | |
Tamper Evident, Flip Top |
Polypropylene | |
45 mL | |
1.5 oz. | |
Specimen Container |