missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CapMat, for use with 96-well MASTERBLOCK™ Microplates
GREENER_CHOICE

Product Code. 10708264
Click to view available options
Pre-slit:
No
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10708264

Brand: Greiner Bio-One 381061

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sealers for microplates

  • For use with 96-well MASTERBLOCK Microplates
TRUSTED_SUSTAINABILITY

Specifications

Color Translucent
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens; resistant to DMSO; non-cytotoxic
Material EVA
Sterility Sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Case 50
No. per Pack 1
Pre-slit No
Pierceable No
Temperature Range (Metric) -60°C to +65°C
Well Shape Round
Plate Compatibility 96-well MASTERBLOCK (0.5 and 1 mL), Standard 96-well Polypropylene Microplates, 96-well F-, U-, and μ Clear-bottom Polystyrene Microplates
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.