Learn More
Capping Reagent, Honeywell Burdick & Jackson™
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 84/16 THF/NMI (v/v)
Brand: Honeywell Fluka BR650-4x2.5L
Additional Details : CAS Number : 109-99-9 Weight : 0.89580kg
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
616-47-7,109-99-9 | |
66°C.The physical data is that of the main component. | |
-15°C | |
Liquid, clear |
84.00% | |
4 x 2.5 L | |
Glass Bottle | |
Capping reagents |