missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
£58.20 - £944.04
Chemical Identifiers
CAS | 502-44-3 |
---|---|
MDL Number | MFCD00003267 |
InChI Key | PAPBSGBWRJIAAV-UHFFFAOYSA-N |
Synonym | 6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane |
PubChem CID | 10401 |
ChEBI | CHEBI:17915 |
IUPAC Name | oxepan-2-one |
SMILES | C1CCC(=O)OCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10092080
|
Thermo Scientific Acros
173442500 |
250 g | Glass bottle |
£58.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10742601
|
Thermo Scientific Acros
173440010 |
1 kg | Glass bottle |
£159.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173713
|
Thermo Scientific Acros
173440025 |
2.5 kg | Glass bottle |
£321.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10388539
|
Thermo Scientific Acros
173440100 |
10 kg | Plastic drum | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
502-44-3 | |
PAPBSGBWRJIAAV-UHFFFAOYSA-N | |
10401 | |
oxepan-2-one |
MFCD00003267 | |
6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane | |
CHEBI:17915 | |
C1CCC(=O)OCC1 |
Specifications
502-44-3 | |
Colorless | |
235°C | |
Authentic | |
C6H10O2 | |
MFCD00003267 | |
1.03 | |
Solubility in water: >1000mg/l (20°C). Other solubilities: soluble in ethanol,acetone,diethyl ether | |
C1CCC(=O)OCC1 | |
114.14 | |
CHEBI:17915 | |
99% | |
ε-Caprolactone monomer |
-1°C | |
1.0300g/mL | |
109°C | |
98.5% min. (GC) | |
1.4620 to 1.464 | |
17,II,29 | |
6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane | |
PAPBSGBWRJIAAV-UHFFFAOYSA-N | |
oxepan-2-one | |
10401 | |
114.14 | |
Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-938-1
RUO – Research Use Only