missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capsazepine, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Descripción
Selective vanilloid receptor antagonist (Ki = 3.2 μM). Inhibits carrageenan inflammation-induced hyperalgesic responses in the rat. Also activates amiloride-sensitive epithelial Na+ channel ENaCδ.
Especificaciones
Quantity | 10 mg |
Formula Weight | 376.9 |
Percent Purity | >99% |
Chemical Name or Material | Capsazepine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'